Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008052.3 | NP_032078.2 | 4 - 5 | 1876 | 115 | |
GenBank mRNA | AB015422.1 | - | 3 - 4 | 1479 | 115 | |
AK088630.1 | - | 4 - 5 | 1832 | 115 | ||
AK169455.1 | - | 4 - 5 | 1877 | 115 | ||
AK220514.1 | - | 3 - 4 | 1457 | 115 | ||
BC053055.1 | - | 4 - 5 | 1022 | 5685525 | 115 | |
BC065075.1 | - | 5 - 6 | 1658 | 115 | ||
U38252.1 | - | 2 - 3 | 1061 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |