Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lynx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011838.4 | NP_035968.1 | 1 - 2 | 92 | 94 | |
GenBank mRNA | AF141377.1 | - | 1 - 2 | 66 | 94 | |
AK161907.1 | - | 1 - 2 | 122 | 94 | ||
AK162920.1 | - | 1 - 2 | 115 | 94 | ||
AK165295.1 | - | 1 - 2 | 96 | 94 | ||
AK165314.1 | - | 1 - 2 | 96 | 94 | ||
BC037541.1 | - | 1 - 2 | 139 | 5325123 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |