Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019516.3 | NP_062389.1 | 5 - 6 | 979 | 95 | |
GenBank mRNA | AF223223.1 | - | 6 - 7 | 673 | 95 | |
AF244978.1 | - | 6 - 7 | 681 | 95 | ||
AF244979.1 | - | 5 - 6 | 726 | 95 | ||
AK085660.1 | - | 5 - 6 | 982 | 95 | ||
AK136526.1 | - | 5 - 6 | 980 | 95 | ||
BC030890.1 | - | 5 - 6 | 663 | 4485119 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |