Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009965.2 | NP_034095.1 | 2 - 3 | 104 | 66 | |
GenBank mRNA | AJ239052.1 | - | 2 - 3 | 104 | 66 | |
BC107342.2 | - | 2 - 3 | 104 | 40055280 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |