Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011715.2 | NP_035845.1 | 11 - 12 | 1349 | 72 | |
GenBank mRNA | AF020055.1 | - | 11 - 12 | 1349 | 72 | |
AK004644.1 | - | 11 - 12 | 1457 | 72 | ||
AK004858.1 | - | 11 - 12 | 1420 | 72 | ||
AK151146.1 | - | 11 - 12 | 1346 | 72 | ||
AK153338.1 | - | 11 - 12 | 1360 | 72 | ||
AK156300.1 | - | 11 - 12 | 1386 | 72 | ||
AK165225.1 | - | 11 - 12 | 1319 | 72 | ||
AK165323.1 | - | 11 - 12 | 1320 | 72 | ||
AK167241.1 | - | 11 - 12 | 1375 | 72 | ||
AK167275.1 | - | 11 - 12 | 1417 | 72 | ||
AK167813.1 | - | 11 - 12 | 1347 | 72 | ||
AK169835.1 | - | 11 - 12 | 1443 | 72 | ||
AK169893.1 | - | 11 - 12 | 1379 | 72 | ||
AK170379.1 | - | 11 - 12 | 1359 | 72 | ||
AK177180.1 | - | 3 - 4 | 346 | 72 | ||
AK194295.1 | - | 2 - 3 | 195 | 72 | ||
AK194906.1 | - | 2 - 3 | 191 | 72 | ||
AK206614.1 | - | 2 - 3 | 121 | 72 | ||
BC049117.1 | - | 11 - 12 | 1441 | 72 | ||
BC119358.1 | - | 11 - 12 | 1305 | 8734107 | 72 | |
BC125295.1 | - | 11 - 12 | 1305 | 40129810 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |