Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195083.1 | NP_001182012.1 | 3 - 4 | 628 | 88 | |
NM_001195130.1 | NP_001182059.1 | 11 - 12 | 2028 | 88 | ||
NM_018774.4 | NP_061244.1 | 11 - 12 | 2021 | 88 | ||
GenBank mRNA | AB062362.1 | - | 3 - 4 | 614 | 88 | |
AF060076.1 | - | 4 - 5 | 607 | 88 | ||
BC051657.1 | - | 3 - 4 | 88 | |||
BC057571.1 | - | 11 - 12 | 1964 | 6410302 | 88 | |
BC071246.1 | - | 3 - 4 | 609 | 88 | ||
U81491.1 | - | 10 - 11 | 1918 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |