Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159905.1 | NP_001153377.1 | 4 - 5 | 363 | 86 | |
NM_001159906.1 | NP_001153378.1 | 4 - 5 | 360 | 86 | ||
NM_026846.3 | NP_081122.2 | 3 - 4 | 397 | 86 | ||
GenBank mRNA | AK004332.1 | - | 3 - 4 | 387 | 86 | |
AK151430.1 | - | 3 - 4 | 399 | 86 | ||
BC011495.1 | - | 3 - 4 | 422 | 4196340 | 86 | |
CX217579.1 | - | 4 - 5 | 366 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |