Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrcc6bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159559.1 | NP_001153031.1 | 2 - 3 | 317 | 118 | |
NM_026858.3 | NP_081134.2 | 2 - 3 | 317 | 118 | ||
GenBank mRNA | AK004405.1 | - | 2 - 3 | 297 | 118 | |
AK010471.1 | - | 2 - 3 | 118 | |||
AK039002.1 | - | 2 - 3 | 298 | 118 | ||
BC030630.1 | - | 2 - 3 | 279 | 1196003 | 118 | |
BG801921.1 | - | 2 - 3 | 317 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |