Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ciz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252534.1 | NP_001239463.1 | 14 - 15 | 2305 | 60 | |
NM_001252536.1 | NP_001239465.1 | 15 - 16 | 2017 | 60 | ||
NM_001252537.1 | NP_001239466.1 | 14 - 15 | 2278 | 60 | ||
NM_001252538.1 | NP_001239467.1 | 14 - 15 | 1968 | 60 | ||
NM_028412.2 | NP_082688.1 | 13 - 14 | 2082 | 60 | ||
GenBank mRNA | AJ575057.1 | - | 12 - 13 | 1589 | 60 | |
AK013708.1 | - | 1 - 2 | 61 | 60 | ||
AK089986.1 | - | 14 - 15 | 2281 | 60 | ||
AK137961.1 | - | 14 - 15 | 1884 | 60 | ||
AK159103.1 | - | 14 - 15 | 2283 | 60 | ||
AK165687.1 | - | 15 - 16 | 1971 | 60 | ||
AK194903.1 | - | 1 - 2 | 36 | 60 | ||
BC018483.1 | - | 13 - 14 | 2046 | 3597692 | 60 | |
BC036730.1 | - | 1 - 2 | 1936 | 60 | ||
BC072624.1 | - | 3168 | 6825309 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |