Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam98c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146023.1 | NP_001139495.1 | 3 - 4 | 366 | 86 | |
GenBank mRNA | AK003496.1 | - | 2 - 3 | 293 | 86 | |
AK080790.1 | - | 2 - 3 | 458 | 86 | ||
AK138395.1 | - | 3 - 4 | 340 | 86 | ||
AK166139.1 | - | 3 - 4 | 336 | 86 | ||
BC058765.1 | - | 2 - 3 | 423 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |