Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hhatl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029095.2 | NP_083371.2 | 2 - 3 | 361 | 97 | |
NR_027967.1 | 2 - 3 | 241 | 97 | |||
GenBank mRNA | AK003605.1 | - | 2 - 3 | 238 | 97 | |
AK157924.1 | - | 2 - 3 | 159 | 97 | ||
BC024464.1 | - | 2 - 3 | 217 | 4975643 | 97 | |
BY703867.1 | - | 2 - 3 | 238 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |