Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053159.3 | NP_444389.2 | 4 - 5 | 616 | 76 | |
GenBank mRNA | AB049633.1 | - | 4 - 5 | 497 | 76 | |
AK008592.1 | - | 4 - 5 | 646 | 76 | ||
AK054185.1 | - | 1 - 2 | 51 | 76 | ||
AK077857.1 | - | 4 - 5 | 616 | 76 | ||
AK145687.1 | - | 4 - 5 | 646 | 76 | ||
AK169272.1 | - | 4 - 5 | 617 | 76 | ||
BC011128.1 | - | 4 - 5 | 585 | 4187019 | 76 | |
BC016586.1 | - | 4 - 5 | 489 | 2616397 | 76 | |
BC034877.1 | - | 4 - 5 | 582 | 1547459 | 76 | |
KC508086.1 | - | 4 - 5 | 593 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |