Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Puf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164600.1 | NP_001158072.1 | 2 - 3 | 170 | 139 | |
NM_028364.2 | NP_082640.2 | 2 - 3 | 200 | 139 | ||
NM_133691.5 | NP_598452.2 | 2 - 3 | 200 | 139 | ||
GenBank mRNA | AK012434.1 | - | 2 - 3 | 140 | 139 | |
AK075960.1 | - | 2 - 3 | 168 | 139 | ||
AK146534.1 | - | 2 - 3 | 170 | 139 | ||
AK149632.1 | - | 2 - 3 | 166 | 139 | ||
AK151526.1 | - | 2 - 3 | 164 | 139 | ||
AK168789.1 | - | 2 - 3 | 167 | 139 | ||
BC010601.1 | - | 2 - 3 | 170 | 3257153 | 139 | |
BY296547.1 | - | 2 - 3 | 201 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |