Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam98a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133747.2 | NP_598508.2 | 3 - 4 | 488 | 85 | |
GenBank mRNA | AK012997.1 | - | 3 - 4 | 422 | 85 | |
AK146215.1 | - | 3 - 4 | 432 | 85 | ||
AK146761.1 | - | 3 - 4 | 454 | 85 | ||
AK146955.1 | - | 3 - 4 | 401 | 85 | ||
AK167224.1 | - | 3 - 4 | 372 | 85 | ||
BC004626.1 | - | 3 - 4 | 385 | 3585698 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |