Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020622.2 | NP_065647.1 | 5 - 6 | 496 | 90 | |
GenBank mRNA | AF360358.1 | - | 5 - 6 | 497 | 90 | |
AF494380.1 | - | 5 - 6 | 401 | 90 | ||
AK018571.1 | - | 5 - 6 | 466 | 90 | ||
BC048949.1 | - | 5 - 6 | 443 | 6474753 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |