Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026240.2 | NP_080516.2 | 3 - 4 | 457 | 93 | |
GenBank mRNA | AK018551.1 | - | 4 - 5 | 707 | 93 | |
AK018691.1 | - | 3 - 4 | 443 | 93 | ||
AK038289.1 | - | 2 - 3 | 498 | 93 | ||
AK161329.1 | - | 3 - 4 | 390 | 93 | ||
AK164050.1 | - | 3 - 4 | 442 | 93 | ||
BC057978.1 | - | 3 - 4 | 432 | 5044170 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |