Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100019389.mm.1 |
NM_001285826.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019389.mm.1 |
NM_001285827.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019389.mm.1 |
NM_027351.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019389.mm.1 |
NM_027351.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019389.mm.1 |
NM_027374.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019389.mm.1 |
NM_027374.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019389.mm.1 |
TC0100002382.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019385.mm.1 |
NM_001285826.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019385.mm.1 |
NM_001285827.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019385.mm.1 |
NM_027351.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019385.mm.1 |
NM_027351.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019385.mm.1 |
NM_027374.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019385.mm.1 |
NM_027374.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019385.mm.1 |
TC0100002382.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019386.mm.1 |
NM_001285826.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019386.mm.1 |
NM_001285827.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019386.mm.1 |
NM_027351.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019386.mm.1 |
NM_027351.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019386.mm.1 |
NM_027374.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019386.mm.1 |
NM_027374.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019386.mm.1 |
TC0100002382.mm.1 |
MTA TC |