Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK018216.1 | - | 571 | 113 | ||
BC094367.1 | - | 220 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried |