Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2005685 |
AK164311.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2005685 |
AK088316.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2005685 |
AK006967.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2005685 |
AK010877.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2005680 |
BC029746.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2005680 |
BC031900.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2005680 |
AK010877.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2005680 |
NM_029665.3 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
468960 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
192777 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
418484 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
926447 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
408346 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
57521 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
587629 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
5580 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
626212 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
242298 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
97747 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
876190 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
210930 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
139418 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
616996 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
384234 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
81496 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
899149 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
355471 |
NM_178199.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
354524 |
AK017118.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
354524 |
AK042409.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
354524 |
AK076332.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
354524 |
BC014719.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
354524 |
NM_028965.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
284365 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
374986 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
466024 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
322845 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
772095 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
495437 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
581062 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
292256 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
535794 |
AK006642.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
535794 |
XM_001475366.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
535794 |
XM_001478509.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
8372 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
507968 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
593461 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900827 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
306077 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
533300 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
675224 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
177934 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
542174 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
725262 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
769061 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
391364 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
476217 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
884771 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
700797 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
649355 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
154110 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
255115 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
645765 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
154719 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
510044 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
671098 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
958749 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
715502 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
301896 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
265277 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
522326 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
592810 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
10530 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
91463 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
966936 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
642413 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
258297 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
278436 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
626559 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
508341 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
945961 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
514888 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
29124 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
73671 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
892729 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
685307 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
697522 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
904756 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
183433 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
257010 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
183058 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
327494 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
879318 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
135940 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
703330 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
387879 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
55929 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
848879 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
335717 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
501265 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
500399 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
913310 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
482070 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
544442 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
2422 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
10381 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
21980 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
136282 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
68926 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
66824 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
75195 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
98453 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
145146 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
52773 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
74329 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
49746 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
155372 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
172933 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P171665 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P146085 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P143607 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2005680 |
BC029746.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2005680 |
BC031900.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2005680 |
AK010877.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2005680 |
NM_029665.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P141432 |
BC029746.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P141432 |
BC031900.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P141432 |
AK087793.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P141432 |
AK044487.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P141432 |
NM_029665.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P401304 |
BC031900.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P401304 |
AK010877.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P401304 |
NM_029665.3 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07986197 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07986584 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07986214 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P401304 |
BC031900.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P401304 |
AK010877.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P401304 |
NM_029665.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P499934 |
BC029746.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P499934 |
BC031900.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P499934 |
AK087373.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P499934 |
AK087793.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P499934 |
NM_029665.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P625461 |
AK164311.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P625461 |
AK088316.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P625461 |
AK006967.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P141432 |
BC029746.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P141432 |
BC031900.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P141432 |
AK087793.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P141432 |
AK044487.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P141432 |
NM_029665.3 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P139650 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P07986584 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P07986209 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10823826 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10823815 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10823817 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10823809 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10745922 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10745927 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301498 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301479 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301484 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301525 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301511 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301515 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301523 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301518 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301499 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301483 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301532 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301494 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301517 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301505 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301474 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301486 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301476 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301519 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301513 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301477 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301527 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301502 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301472 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301496 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301500 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301481 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301521 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301535 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301504 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301489 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301506 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301528 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301509 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301524 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301501 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301533 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29301503 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29301474 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29301476 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32904462 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10481421 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29301479 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29301475 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29301472 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29301481 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29301469 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29301471 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29301477 |
- |
- |