Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3c5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290484.1 | NP_001277413.1 | 2 - 3 | 670 | 78 | |
NM_148928.3 | NP_683730.1 | 2 - 3 | 670 | 78 | ||
GenBank mRNA | AK045419.1 | - | 2 - 3 | 262 | 78 | |
AK147014.1 | - | 2 - 3 | 648 | 78 | ||
BC027055.1 | - | 2 - 3 | 637 | 5360397 | 78 | |
BC027247.1 | - | 2 - 3 | 580 | 2631223 | 78 | |
BC039171.1 | - | 2 - 3 | 591 | 4500799 | 78 | |
BQ179286.1 | - | 2 - 3 | 670 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |