Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baz1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011714.2 | NP_035844.2 | 7 - 8 | 2929 | 67 | |
GenBank mRNA | AF084480.1 | - | 7 - 8 | 2888 | 67 | |
AK037737.1 | - | 1 - 2 | 137 | 67 | ||
BC141399.1 | - | 7 - 8 | 2832 | 9056012 | 67 | |
BC171975.1 | - | 7 - 8 | 2832 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |