Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015792.1 | NP_056607.1 | 12 - 13 | 2050 | 79 | |
GenBank mRNA | AF184275.1 | - | 10 - 11 | 1544 | 79 | |
AK083118.1 | - | 12 - 13 | 2395 | 79 | ||
AK147829.1 | - | 12 - 13 | 2034 | 79 | ||
AK154357.1 | - | 12 - 13 | 2073 | 79 | ||
AK158937.1 | - | 12 - 13 | 2101 | 79 | ||
AK190454.1 | - | 2 - 3 | 132 | 79 | ||
AK192824.1 | - | 3 - 4 | 259 | 79 | ||
AK204859.1 | - | 2 - 3 | 131 | 79 | ||
AK216886.1 | - | 2 - 3 | 131 | 79 | ||
BC004036.1 | - | 9 - 10 | 1136 | 79 | ||
BC031393.1 | - | 12 - 13 | 2050 | 4948804 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |