Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145524.3 | NP_663499.2 | 1 - 2 | 232 | 112 | |
GenBank mRNA | AK039461.1 | - | 1 - 2 | 199 | 112 | |
AK042669.1 | - | 1 - 2 | 210 | 112 | ||
AK087927.1 | - | 1 - 2 | 161 | 111 | ||
BC004636.1 | - | 1 - 2 | 153 | 3585719 | 112 | |
DQ093357.1 | - | 1 - 2 | 209 | 112 | ||
DQ093358.1 | - | 1 - 2 | 209 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |