Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146089.2 | NP_666201.1 | 1 - 2 | 60 | 67 | |
GenBank mRNA | AK076912.1 | - | 1 - 2 | 40 | 67 | |
AK146923.1 | - | 1 - 2 | 57 | 67 | ||
AK147599.1 | - | 1 - 2 | 67 | 67 | ||
BC024400.1 | - | 1 - 2 | 60 | 5026575 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |