Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172422.3 | NP_766010.1 | 5 - 6 | 1228 | 93 | |
GenBank mRNA | AK035322.1 | - | 5 - 6 | 1228 | 93 | |
AK084971.1 | - | 5 - 6 | 1203 | 93 | ||
AK148224.1 | - | 1212 | 93 | |||
AK153916.1 | - | 4 - 5 | 1171 | 93 | ||
AK167714.1 | - | 4 - 5 | 1484 | 93 | ||
BC008271.1 | - | 4 - 5 | 1169 | 3603187 | 93 | |
BC057208.1 | - | 4 - 5 | 1162 | 5328766 | 93 | |
BC080813.1 | - | 4 - 5 | 1168 | 6394260 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |