Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700030147.mm.1 |
NM_001113471.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030147.mm.1 |
NM_001113471.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030147.mm.1 |
NM_175640.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030147.mm.1 |
NM_175640.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030147.mm.1 |
TC0700003567.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700030146.mm.1 |
NM_001113471.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030146.mm.1 |
NM_001113471.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030146.mm.1 |
NM_175640.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030146.mm.1 |
NM_175640.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030146.mm.1 |
TC0700003567.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700030149.mm.1 |
NM_175640.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030149.mm.1 |
NM_175640.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030149.mm.1 |
TC0700003567.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700030150.mm.1 |
NM_001113471.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030150.mm.1 |
NM_001113471.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030150.mm.1 |
NM_175640.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030150.mm.1 |
NM_175640.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030150.mm.1 |
TC0700003567.mm.1 |
MTA TC |