Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400015740.mm.1 |
NM_080728.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015740.mm.1 |
NM_080728.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015740.mm.1 |
TC1400002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015741.mm.1 |
NM_080728.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015741.mm.1 |
NM_080728.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015741.mm.1 |
TC1400002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015742.mm.1 |
NM_080728.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015742.mm.1 |
NM_080728.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015742.mm.1 |
TC1400002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015744.mm.1 |
NM_080728.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015744.mm.1 |
NM_080728.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015744.mm.1 |
TC1400002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015749.mm.1 |
NM_080728.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015749.mm.1 |
NM_080728.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015749.mm.1 |
TC1400002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015743.mm.1 |
NM_080728.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015743.mm.1 |
NM_080728.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015743.mm.1 |
TC1400002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015748.mm.1 |
NM_080728.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015748.mm.1 |
NM_080728.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015748.mm.1 |
TC1400002075.mm.1 |
MTA TC |