Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnali1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175223.4 | NP_780432.1 | 4 - 5 | 630 | 71 | |
GenBank mRNA | AK077109.1 | - | 4 - 5 | 631 | 71 | |
BC118619.1 | - | 4 - 5 | 608 | 40107304 | 71 | |
BC122552.1 | - | 4 - 5 | 605 | 40107303 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |