Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177615.3 | NP_808283.2 | 7 - 8 | 1235 | 104 | |
GenBank mRNA | AK052641.1 | - | 1174 | 104 | ||
BC089587.1 | - | 7 - 8 | 1172 | 6757541 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |