Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P671794 |
AK166080.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P671794 |
BC052067.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P671794 |
AK141580.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P671794 |
AK165919.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P671794 |
AK079413.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P671794 |
AK166080.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P671794 |
BC052067.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P671794 |
AK141580.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P671794 |
AK165919.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P671794 |
AK079413.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
316156 |
AF229432.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
316156 |
AF229433.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
316156 |
AF229434.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
316156 |
AK020762.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
316156 |
AK081878.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
316156 |
AY157832.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
316156 |
AY157833.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
316156 |
NM_009400.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
316156 |
NM_021985.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
316156 |
U82534.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
893161 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
654456 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
889621 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
631858 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
431872 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
836562 |
AF276425.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
836562 |
AK041590.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
836562 |
AK083722.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
836562 |
BC066066.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
836562 |
BC080278.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
836562 |
NM_022723.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
108549 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
322662 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
183567 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
497506 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
760461 |
AK019121.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
760461 |
AK019124.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
760461 |
AK145685.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
447122 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
885621 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
826462 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
494948 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
309787 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
594682 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
923704 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
64718 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
970039 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
94303 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
96762 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
61366 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P168429 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P178402 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P671794 |
AK166080.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P671794 |
BC052067.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P671794 |
AK141580.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P671794 |
AK165919.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P671794 |
AK079413.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P671794 |
AK166080.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P671794 |
BC052067.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P671794 |
AK141580.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P671794 |
AK165919.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P671794 |
AK079413.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06882157 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P168429 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06882116 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P178402 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P671794 |
AK166080.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P671794 |
BC052067.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P671794 |
AK141580.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P671794 |
AK165919.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P671794 |
AK079413.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P671794 |
AK166080.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P671794 |
BC052067.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P671794 |
AK141580.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P671794 |
AK165919.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P671794 |
AK079413.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P132937 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P168429 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P178402 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07455273 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07689530 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07455277 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07455265 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07689531 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07455275 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07689527 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726830 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726690 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726767 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726702 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726777 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726851 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726697 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726822 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726866 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726843 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726728 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726827 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726826 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726764 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726856 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726723 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726756 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726721 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726812 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726730 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726841 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726716 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726799 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726802 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726779 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726801 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726772 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726705 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726703 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726761 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726805 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726837 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726807 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726775 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726829 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726784 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726743 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726698 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726760 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726694 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726685 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726710 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726747 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726759 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726783 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726740 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726874 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726719 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726744 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726795 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726738 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726872 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726737 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726831 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726787 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726792 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726819 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726774 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726782 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726691 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726712 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726750 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726865 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726861 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726748 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726847 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726810 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726869 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726858 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726852 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726720 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726762 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726692 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726867 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726816 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726823 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726839 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726804 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726757 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726786 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726835 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726687 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726868 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726704 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726849 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726709 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726797 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726814 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726806 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726754 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726875 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26726863 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26726856 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08013529 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26726882 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26726877 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07815292 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26726860 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26726685 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26726861 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26726875 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26726876 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26726880 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26726853 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07881522 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26726878 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26726682 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26726858 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26726683 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26726865 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26726855 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07715508 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26726681 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07715510 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26726852 |
- |
- |