Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smyd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026796.1 | NP_081072.1 | 4 - 5 | 416 | 103 | |
GenBank mRNA | AK145261.1 | - | 4 - 5 | 421 | 103 | |
AK150857.1 | - | 4 - 5 | 420 | 103 | ||
AK164053.1 | - | 5 - 6 | 926 | 103 | ||
BC023119.1 | - | 4 - 5 | 416 | 4458755 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |