Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178650.3 | NP_848765.2 | 3 - 4 | 295 | 115 | |
GenBank mRNA | AK040487.1 | - | 3 - 4 | 296 | 115 | |
AK075803.1 | - | 3 - 4 | 282 | 115 | ||
AK088119.1 | - | 360 | 115 | |||
AK131177.1 | - | 2 - 3 | 276 | 115 | ||
AK146426.1 | - | 3 - 4 | 115 | |||
AK153836.1 | - | 3 - 4 | 293 | 115 | ||
AK154444.1 | - | 2 - 3 | 273 | 115 | ||
BC094901.1 | - | 2 - 3 | 270 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |