Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027257.1 | NP_081533.1 | 4 - 5 | 367 | 179 | |
GenBank mRNA | AK011703.1 | - | 3 - 4 | 335 | 179 | |
AK207620.1 | - | 2 - 3 | 202 | 179 | ||
AK208217.1 | - | 2 - 3 | 202 | 179 | ||
BC026942.1 | - | 4 - 5 | 367 | 5011571 | 179 | |
DQ158906.1 | - | 4 - 5 | 367 | 179 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |