Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143755.1 | NP_001137227.1 | 5 - 6 | 709 | 105 | |
NM_027164.1 | NP_081440.1 | 4 - 5 | 754 | 105 | ||
GenBank mRNA | AK006931.1 | - | 5 - 6 | 714 | 105 | |
BC050816.1 | - | 4 - 5 | 754 | 6774506 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |