Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038604.1 | NP_001033693.1 | 3 - 4 | 333 | 71 | |
GenBank mRNA | BC104364.1 | - | 3 - 4 | 292 | 40049817 | 71 |
BC104365.1 | - | 3 - 4 | 292 | 40049818 | 71 | |
CN670295.1 | - | 3 - 4 | 295 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |