Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thsd7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164805.1 | NP_001158277.1 | 9 - 10 | 2630 | 63 | |
GenBank mRNA | AK133324.1 | - | 4 - 5 | 710 | 63 | |
AK173072.1 | - | 9 - 10 | 2401 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Mus musculus Probe spans exons |