Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmbim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026617.3 | NP_080893.1 | 5 - 6 | 485 | 89 | |
GenBank mRNA | AK006207.1 | - | 5 - 6 | 486 | 89 | |
AK153442.1 | - | 4 - 5 | 413 | 89 | ||
AK184146.1 | - | 3 - 4 | 250 | 89 | ||
AK208947.1 | - | 3 - 4 | 216 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |