Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sez6l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252566.1 | NP_001239495.1 | 13 - 14 | 2797 | 104 | |
NM_001252567.1 | NP_001239496.1 | 13 - 14 | 2617 | 104 | ||
NM_144926.5 | NP_659175.1 | 13 - 14 | 2797 | 104 | ||
GenBank mRNA | AB206789.1 | - | 13 - 14 | 2398 | 104 | |
AK034088.1 | - | 13 - 14 | 2769 | 104 | ||
BC011475.1 | - | 13 - 14 | 2603 | 4191879 | 104 | |
BC079573.1 | - | 13 - 14 | 2186 | 5686330 | 104 | |
BC096615.1 | - | 13 - 14 | 2709 | 6467632 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |