Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bet1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009748.2 | NP_033878.1 | 2 - 3 | 277 | 146 | |
GenBank mRNA | AF007552.1 | - | 2 - 3 | 265 | 146 | |
AK078796.1 | - | 2 - 3 | 273 | 146 | ||
BC005572.1 | - | 2 - 3 | 277 | 3967968 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |