Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177633.4 | NP_808301.3 | 4 - 5 | 396 | 118 | |
GenBank mRNA | AK147730.1 | - | 4 - 5 | 392 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mus musculus Probe spans exons |