Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190473.1 | NP_001177402.1 | 2 - 3 | 143 | 77 | |
NM_001190474.1 | NP_001177403.2 | 2 - 3 | 195 | 77 | ||
NM_007828.2 | NP_031854.1 | 1 - 2 | 163 | 77 | ||
GenBank mRNA | AB007143.1 | - | 1 - 2 | 76 | 77 | |
AK158299.1 | - | 2 - 3 | 186 | 77 | ||
BC012210.1 | - | 2 - 3 | 161 | 77 | ||
BC023238.1 | - | 2 - 3 | 133 | 77 | ||
BC125443.1 | - | 1 - 2 | 163 | 40129958 | 77 | |
BC137680.1 | - | 1 - 2 | 163 | 8860699 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |