Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029806.1 | NP_084082.1 | 3 - 4 | 381 | 76 | |
GenBank mRNA | AK008654.1 | - | 3 - 4 | 384 | 76 | |
BC107186.2 | - | 3 - 4 | 381 | 40050796 | 76 | |
BC107187.2 | - | 3 - 4 | 381 | 40050800 | 76 | |
BC119485.1 | - | 3 - 4 | 381 | 40050803 | 76 | |
BC119486.1 | - | 3 - 4 | 381 | 40050804 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |