Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133981.2 | NP_598742.1 | 9 - 10 | 1177 | 73 | |
NR_045601.1 | 8 - 9 | 1106 | 73 | |||
GenBank mRNA | AK050335.1 | - | 9 - 10 | 1098 | 73 | |
AK054293.1 | - | 9 - 10 | 1083 | 73 | ||
AK086674.1 | - | 9 - 10 | 1117 | 73 | ||
AK149592.1 | - | 8 - 9 | 1028 | 73 | ||
BC021791.1 | - | 9 - 10 | 1072 | 4459738 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |