Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnfn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028219.1 | NP_082495.1 | 1 - 2 | 64 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |