Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsbg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053178.2 | NP_444408.1 | 2 - 3 | 437 | 84 | |
GenBank mRNA | AB050554.1 | - | 2 - 3 | 437 | 84 | |
AK054103.1 | - | 2 - 3 | 250 | 84 | ||
AK129179.1 | - | 2 - 3 | 233 | 84 | ||
BC057322.1 | - | 2 - 3 | 233 | 6832261 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |