Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252282.1 | NP_001239211.1 | 3 - 4 | 535 | 60 | |
NM_001252287.1 | NP_001239216.1 | 3 - 4 | 611 | 60 | ||
NM_010956.4 | NP_035086.2 | 3 - 4 | 535 | 60 | ||
GenBank mRNA | AK147289.1 | - | 3 - 4 | 536 | 60 | |
AK150009.1 | - | 3 - 4 | 501 | 60 | ||
AK169286.1 | - | 3 - 4 | 527 | 60 | ||
AK220536.1 | - | 3 - 4 | 497 | 60 | ||
BC013670.1 | - | 3 - 4 | 469 | 4188988 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |