Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmiz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005867.1 | NP_001005867.1 | 14 - 15 | 2055 | 83 | |
NM_028601.3 | NP_082877.2 | 14 - 15 | 2151 | 83 | ||
GenBank mRNA | BC024081.1 | - | 13 - 14 | 1901 | 83 | |
BC040745.1 | - | 5 - 6 | 610 | 83 | ||
BC060652.1 | - | 14 - 15 | 1993 | 30354393 | 83 | |
BC063069.1 | - | 6 - 7 | 756 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |