Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015768.2 | NP_056583.1 | 2 - 3 | 296 | 86 | |
GenBank mRNA | AF182064.1 | - | 2 - 3 | 296 | 86 | |
AK015462.1 | - | 2 - 3 | 258 | 86 | ||
BC116911.1 | - | 2 - 3 | 265 | 40126230 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |