Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033253.3 | NP_001028425.1 | 5 - 6 | 792 | 88 | |
NM_001159942.1 | NP_001153414.1 | 6 - 7 | 958 | 88 | ||
GenBank mRNA | AK033676.1 | - | 6 - 7 | 958 | 88 | |
AK034403.1 | - | 6 - 7 | 959 | 88 | ||
AK045140.1 | - | 6 - 7 | 1138 | 88 | ||
AK122468.1 | - | 2 - 3 | 116 | 88 | ||
AK135923.1 | - | 5 - 6 | 794 | 88 | ||
BC076628.1 | - | 3 - 4 | 189 | 88 | ||
BC117965.1 | - | 4 - 5 | 394 | 40098326 | 88 | |
BC117966.1 | - | 4 - 5 | 387 | 40098330 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |