Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acrbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016845.2 | NP_058541.2 | 6 - 7 | 1152 | 67 | |
GenBank mRNA | AK132705.1 | - | 6 - 7 | 1120 | 67 | |
D17573.1 | - | 6 - 7 | 1104 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |